4-(Trifluormethoxy)toluol

Lieferant: MENAI

CF4.25 CF4.5
MENACF4.25EA 0 EUR
MENACF4.25 MENACF4.5
4-(Trifluormethoxy)toluol
4-(Trifluormethoxy)toluol
Formel: C₈H₇F₃O
MDL: MFCD00042413
CAS-Nummer: 706-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR