Bis(2-bromethyl)ether, Sigma-Aldrich®

Lieferant: Merck

95254-10ML-F 95254-50ML-F
SIAL95254-10ML-FEA 139 EUR
SIAL95254-10ML-F SIAL95254-50ML-F
Bis(2-bromethyl)ether, Sigma-Aldrich®
Bis(2-bromethyl)ether
Formel: C₄H₈Br₂O
MDL: MFCD00039196
CAS-Nummer: 5414-19-7
EINECS: 226-504-2
UN: 2810
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR