(Brommethyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

269158-25G 269158-5G
SIAL269158-25GEA 130 EUR
SIAL269158-25G SIAL269158-5G
(Brommethyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(Bromomethyl)triphenylphosphonium bromide
Formel: C₁₉H₁₇Br₂P
Molecular Weight: 436,1200 g/mol
MDL: MFCD00011864
CAS-Nummer: 1034-49-7
EINECS: 213-857-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR