Tetrakis(decyl)ammoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrakis(decyl)ammonium bromide, IPC-TDA-Br, Tetra(decyl)ammonium bromide

87580-10G 87580-50G
SIAL87580-10GEA 167 EUR
SIAL87580-10G SIAL87580-50G
Tetrakis(decyl)ammoniumbromid, Sigma-Aldrich®
Tetrakis(decyl)ammoniumbromid
Formel: C₄₀H₈₄BrN
Molecular Weight: 659.0057 g/mol
MDL: MFCD00043166
CAS-Nummer: 14937-42-9
EINECS: 239-013-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR