5-(Aminomethyl)-2-brompyridin

Lieferant: COMBI-BLOCKS

OT-8105-1G OT-8105-250M
COBBOT-8105-1GEA 0 EUR
COBBOT-8105-1G COBBOT-8105-250M
5-(Aminomethyl)-2-brompyridin
5-(Aminomethyl)-2-brompyridin
Formel: C₆H₇BrN₂
MDL: MFCD06738744
CAS-Nummer: 120740-10-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR