5-Brom-2-methoxy-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7161-1G PY-7161-5G
COBBPY-7161-1GEA 38.2 EUR
COBBPY-7161-1G COBBPY-7161-5G
5-Brom-2-methoxy-3-nitropyridin
5-Brom-2-methoxy-3-nitropyridin
Formel: C₆H₅BrN₂O₃
Molecular Weight: 233,0200 g/mol
MDL: MFCD07374968
CAS-Nummer: 152684-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR