5-(Aminomethyl)-2-chlorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

536008-25G 536008-5G
SIAL536008-25GEA 390 EUR
SIAL536008-25G SIAL536008-5G
5-(Aminomethyl)-2-chlorpyridin, Sigma-Aldrich®
5-(Aminomethyl)-2-chlorpyridin
Formel: C₆H₇ClN₂
MDL: MFCD00673153
CAS-Nummer: 97004-04-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR