7-Ethoxycumarin ≥99%

Lieferant: Thermo Scientific

Synonyme: 7-Ethoxy-2H-chromen-2-one

L14116.MC L14116.ME
L14116.MCEA 42.4 EUR
L14116.MC L14116.ME
7-Ethoxycumarin ≥99%
7-Ethoxycumarin

Purity: 99% CAS No.: 31005-02-4 Molecular Formula: C11H10O3 Formula Weight: 190.20 Melting Point: 88-91° Beilstein Reference: 146200

Formel: C₁₁H₁₀O₃
Molecular Weight: 190,20 g/mol
Schmelzpunkt: 88…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006877
CAS-Nummer: 31005-02-4
EINECS: 250-429-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR