7-Ethoxycumarin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-Ethoxy-2H-chromen-2-one

M-Clarity™ Qualitätsstufe = MQ200

E1379-100MG E1379-1G E1379-25MG E1379-500MG
SIALE1379-100MGEA 57.4 EUR
SIALE1379-100MG SIALE1379-1G SIALE1379-25MG SIALE1379-500MG
7-Ethoxycumarin, Sigma-Aldrich®
7-Ethoxycumarin
Formel: C₁₁H₁₀O₃
Molecular Weight: 190,20 g/mol
MDL: MFCD00006877
CAS-Nummer: 31005-02-4
EINECS: 250-429-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR