(4-Fluorphenoxy)acetonitril

Lieferant: BIONET RESEARCH

7C-016.500 7C-016.1 7C-016.5 7C-016.10
BIOT7C-016.500EA 0 EUR
BIOT7C-016.500 BIOT7C-016.1 BIOT7C-016.5 BIOT7C-016.10
(4-Fluorphenoxy)acetonitril
(4-Fluorphenoxy)acetonitril
Formel: C₈H₆FNO
CAS-Nummer: 24115-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR