α-Fluorphenylessigsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Fluoro-2-phenylacetic acid, alpha-Fluorophenylacetic acid
009226-1G
009226-5G
FLUO009226-1GEA
149
EUR
FLUO009226-1G
FLUO009226-5G
α-Fluorphenylessigsäure
α-Fluorphenylessigsäure
Formel:
C₈H₇FO₂ Molecular Weight: 154.14 g/mol |
MDL:
MFCD00004221 CAS-Nummer: 1578-63-8 EINECS: 216-420-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...