Isoindolin

Lieferant: FLUOROCHEM

24774-5G 024774-5G
FLUO24774-5GEA 0 EUR
FLUO24774-5G FLUO024774-5G
Isoindolin
Isoindolin
Formel: C₈H₉N
MDL: MFCD00605324
CAS-Nummer: 496-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR