Ethyl-4,4,4-trifluorbutyrat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 4,4,4-trifluorobutanoate, Ethyl 4,4,4-trifluorobutyrate

001498-5G 001498-1G
FLUO001498-5GEA 110 EUR
FLUO001498-5G FLUO001498-1G
Ethyl-4,4,4-trifluorbutyrat
Ethyl-4,4,4-trifluorbutyrat
Formel: C₆H₉F₃O₂
Molecular Weight: 170,13 g/mol
MDL: MFCD00041398
CAS-Nummer: 371-26-6
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR