Perfluor-1-butansulfonsäure

Lieferant: FLUOROCHEM

Synonyme: Perfluoro-1-butanesulphonic acid, Nonafluoro-1-butanesulphonic acid

009307-5G 009307-25G
FLUO009307-5GEA 117 EUR
FLUO009307-5G FLUO009307-25G
Perfluor-1-butansulfonsäure
Perfluor-1-butansulfonsäure
Formel: C₄HF₉O₃S
MDL: MFCD01320794
CAS-Nummer: 375-73-5
EINECS: 206-793-1
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR