Perfluor-1-butansulfonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Perfluoro-1-butanesulphonic acid, Nonafluoro-1-butanesulphonic acid

M-Clarity™ Qualitätsstufe = MQ100

562629-25G 562629-5G
SIAL562629-25GEA 263 EUR
SIAL562629-25G SIAL562629-5G
Perfluor-1-butansulfonsäure, Sigma-Aldrich®
Perfluor-1-butansulfonsäure
Formel: C₄HF₉O₃S
MDL: MFCD01320794
CAS-Nummer: 375-73-5
EINECS: 206-793-1
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR