Lithiumperfluor-1-butansulfonat

Lieferant: FLUOROCHEM

010328-25G 010328-100G
FLUO010328-25GEA 284 EUR
FLUO010328-25G FLUO010328-100G
Lithiumperfluor-1-butansulfonat
Lithiumperfluor-1-butansulfonat
Formel: C₄F₉LiO₃S
MDL: MFCD03094160
CAS-Nummer: 131651-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR