Thuliumtriacetat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Fisher Scientific
Achtung

14582.04 14582.09
14582.04EA 134 EUR
14582.04 14582.09
Thuliumtriacetat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Thuliumtriacetat Hydrat

Purity: 99.9% CAS No.: 314041-04-8 Molecular Formula: Tm(OOCCH3)3.xH2O Formula Weight: 346.07(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic MDL No.: MFCD00013046

Formel: C₆H₉O₆Tm·H₂O
Molecular Weight: 346.07 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013046
CAS-Nummer: 314041-04-8
EINECS: 254-326-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR