Thuliumtriacetat Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

367702-1G 367702-5G
SIAL367702-1GEA 69.2 EUR
SIAL367702-1G SIAL367702-5G
Thuliumtriacetat Hydrat, Sigma-Aldrich®
Thuliumtriacetat Hydrat
Formel: C₆H₉O₆Tm·H₂O
Molecular Weight: 346.07 g/mol (anhy)
MDL: MFCD00013046
CAS-Nummer: 314041-04-8
EINECS: 254-326-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR