Tetraethylorthocarbonat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A17835.06
A17835.14
A17835.22
A17835.06EA
39.5
EUR
A17835.06
A17835.14
A17835.22
Tetraethylorthocarbonat ≥98%
Tetraethylorthocarbonat
Purity: 98+%
CAS No.: 78-09-1
Molecular Formula: C9H20O4
Formula Weight: 192.26
Storage Temperature: 2°C to 8°C
Flash Point: 52°(125°F)
Refractive Index: 1.3930
Boiling Point: 158-161°
Beilstein Reference: 1747401
UN No.: 3272
MDL No.: MFCD00009221
Formel:
C₉H₂₀O₄ Molecular Weight: 192,26 g/mol Siedepunkt: 158…161 °C Dichte: 0,917 Flammpunkt: 52 °C (125 °F) |
MDL:
MFCD00009221 CAS-Nummer: 78-09-1 EINECS: 201-082-2 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...