Cyclopentancarbonitril 98
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A17710.03
A17710.06
A17710.14
A17710.03EA
39.3
EUR
A17710.03
A17710.06
A17710.14
Cyclopentancarbonitril 98
Cyclopentancarbonitril
Purity: 98%
CAS No.: 4254-02-8
Molecular Formula: C6H9N
Formula Weight: 95.14
Flash Point: 56°(133°F)
Refractive Index: 1.4410
Melting Point: -76°
Boiling Point: 67-68°/10mm
Beilstein Reference: 1098717
UN No.: 1992
MDL No.: MFCD00013741
Formel:
C₆H₉N Molecular Weight: 95.14 g/mol Siedepunkt: 67…68 °C (10 mmHg) Schmelzpunkt: –76 °C Dichte: 0.912 Flammpunkt: 56 °C (133 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013741 CAS-Nummer: 4254-02-8 EINECS: 224-229-2 UN: 1992 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...