(4-Methylbenzyl)triphenylphosphoniumbromid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
L01562.06
L01562.14
L01562.06EA
33.3
EUR
L01562.06
L01562.14
(4-Methylbenzyl)triphenylphosphoniumbromid ≥98%
(4-Methylbenzyl)triphenylphosphoniumbromid
Purity: 98+%
CAS No.: 2378-86-1
Molecular Formula: C26H24BrP
Formula Weight: 447.36
Melting Point: 268-270°
Beilstein Reference: 3810292
Formel:
C₂₆H₂₄BrP Molecular Weight: 447,36 g/mol Schmelzpunkt: 268…270 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031580 CAS-Nummer: 2378-86-1 EINECS: 219-159-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...