(4-Chlorbenzyl)triphenylphosphoniumchlorid

Lieferant: CHEMPUR

JFD04036.250 JFD04036.1
CHMPJFD04036.250EA 0 EUR
CHMPJFD04036.250 MAYBJFD04036.1
(4-Chlorbenzyl)triphenylphosphoniumchlorid
(4-Chlorbenzyl)triphenylphosphoniumchlorid
Formel: C₂₅H₂₁Cl₂P
Molecular Weight: 423,32 g/mol
MDL: MFCD00041533
CAS-Nummer: 1530-39-8
EINECS: 216-228-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR