Decafluorbenzhydrol

Lieferant: FLUOROCHEM

Synonyme: Bis(perfluorophenyl)methanol

004610-25G 004610-5G
FLUO004610-25GEA 0 EUR
FLUO004610-25G FLUO004610-5G
Decafluorbenzhydrol
Decafluorbenzhydrol
Formel: C₁₃H₂F₁₀O
Molecular Weight: 364,14 g/mol
MDL: MFCD00000297
CAS-Nummer: 1766-76-3
EINECS: 217-185-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR