Isopropyltriphenylphosphoniumiodid ≥98%
Lieferant: Thermo Scientific
Achtung
A12881.09 A12881.22 A12881.30
A12881.09EA 33.5 EUR
A12881.09 A12881.22 A12881.30
Isopropyltriphenylphosphoniumiodid ≥98%
Isopropyltriphenylphosphoniumiodid
Purity: 98+% CAS No.: 24470-78-8 Molecular Formula: (CH3)2CHP(C6H5)3I Formula Weight: 432.29 Melting Point: 195-198° Beilstein Reference: 3599629
Formel: C₂₁H₂₂IP
Molecular Weight: 432,29 g/mol
Schmelzpunkt: 195…198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031595
CAS-Nummer: 24470-78-8
EINECS: 246-281-5