Isopropyltriphenylphosphoniumiodid

Lieferant: CHEMPUR

CD00116.250 CD00116.1
CHMPCD00116.250EA 0 EUR
CHMPCD00116.250 MAYBCD00116.1
Isopropyltriphenylphosphoniumiodid
Isopropyltriphenylphosphoniumiodid
Formel: C₂₁H₂₂IP
Molecular Weight: 432,29 g/mol
MDL: MFCD00031595
CAS-Nummer: 24470-78-8
EINECS: 246-281-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR