6-Iodnicotinonitril

Lieferant: COMBI-BLOCKS

PY-7009-1G PY-7009-5G
COBBPY-7009-1GEA 0 EUR
COBBPY-7009-1G COBBPY-7009-5G
6-Iodnicotinonitril
6-Iodnicotinonitril
Formel: C₆H₃IN₂
MDL: MFCD08689512
CAS-Nummer: 289470-22-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR