2-Butyl-5-nitrobenzoxazol

Lieferant: BIONET RESEARCH

Synonyme: 2-(But-1-yl)-5-nitro-1,3-benzoxazole

7X-5094.500 7X-5094.1 7X-5094.5 7X-5094.10
BIOT7X-5094.500EA 0 EUR
BIOT7X-5094.500 BIOT7X-5094.1 BIOT7X-5094.5 BIOT7X-5094.10
2-Butyl-5-nitrobenzoxazol
2-Butyl-5-nitrobenzoxazole
Formel: C₁₁H₁₂N₂O₃
MDL: MFCD05663810
CAS-Nummer: 886360-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR