2-Butyl-6-nitrobenzoxazol

Lieferant: BIONET RESEARCH

7X-5093.500 7X-5093.1 7X-5093.5 7X-5093.10
BIOT7X-5093.500EA 0 EUR
BIOT7X-5093.500 BIOT7X-5093.1 BIOT7X-5093.5 BIOT7X-5093.10
2-Butyl-6-nitrobenzoxazol
2-Butyl-6-nitrobenzoxazole
Formel: C₁₁H₁₂N₂O₃
MDL: MFCD05663809
CAS-Nummer: 886360-96-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR