2-Ethyl-6-nitrobenzoxazol

Lieferant: BIONET RESEARCH

1W-0916.500 1W-0916.1 1W-0916.5 1W-0916.10
BIOT1W-0916.500EA 0 EUR
BIOT1W-0916.500 BIOT1W-0916.1 BIOT1W-0916.5 BIOT1W-0916.10
2-Ethyl-6-nitrobenzoxazol
2-Ethyl-6-nitrobenzoxazole
Formel: C₉H₈N₂O₃
CAS-Nummer: 13243-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR