5-(Hydroxymethyl)indol

Lieferant: COMBI-BLOCKS

Synonyme: (1H-Indol-5-yl)methanol

IN-0009-5G IN-0009-1G
COBBIN-0009-5GEA 1100 EUR
COBBIN-0009-5G COBBIN-0009-1G
5-(Hydroxymethyl)indol
5-(Hydroxymethyl)indole
Formel: C₉H₉NO
Molecular Weight: 147,1700 g/mol
MDL: MFCD02179594
CAS-Nummer: 1075-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR