6-(Hydroxymethyl)indol
Lieferant: COMBI-BLOCKS
Synonyme:
(1H-Indol-6-yl)methanol
IN-0016-5G
IN-0016-1G
COBBIN-0016-5GEA
612
EUR
COBBIN-0016-5G
COBBIN-0016-1G
6-(Hydroxymethyl)indol
6-(Hydroxymethyl)indole
Formel:
C₉H₉NO Molecular Weight: 147,1739 g/mol |
MDL:
MFCD02179595 CAS-Nummer: 1075-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...