7-(Hydroxymethyl)indol

Lieferant: COMBI-BLOCKS

Synonyme: (1H-Indol-7-yl)methanol

IN-0021-5G IN-0021-1G
COBBIN-0021-5GEA 560 EUR
COBBIN-0021-5G COBBIN-0021-1G
7-(Hydroxymethyl)indol
7-(Hydroxymethyl)indole
Formel: C₉H₉NO
Molecular Weight: 147,1739 g/mol
MDL: MFCD02179597
CAS-Nummer: 1074-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR