3,3'-Methylendianilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,3'-Methylenedianiline, 3,3'-Diaminodiphenylmethane

M-Clarity™ Qualitätsstufe = MQ100

378267-1G 378267-5G
SIAL378267-1GEA 108 EUR
SIAL378267-1G SIAL378267-5G
3,3'-Methylendianilin, Sigma-Aldrich®
3,3'-Methylendianilin
Formel: C₁₃H₁₄N₂
MDL: MFCD00014777
CAS-Nummer: 19471-12-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR