2-Aminoethyldihydrogenphosphat

Lieferant: FLUOROCHEM

Synonyme: O-Phosphorylethanolamine

010650-25G 010650-100G
FLUO010650-25GEA 65 EUR
FLUO010650-25G FLUO010650-100G
2-Aminoethyldihydrogenphosphat
2-Aminoethyldihydrogenphosphat
Formel: C₂H₈NO₄P
Molecular Weight: 141,0630 g/mol
MDL: MFCD00008178
CAS-Nummer: 1071-23-4
EINECS: 213-988-5
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR