Thulium(III) nitrate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Thulium trinitrate hydrate
14579.04 14579.09 14579.18
14579.04EA 108 EUR
14579.04 14579.09 14579.18
Thulium(III) nitrate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Thulium(III) nitrate hydrate
Purity: 99.9% Molecular Formula: Tm(NO3)3.xH2O (x≈5) Formula Weight: 354.95(anhy) Physical Form: Crystalline Aggregates UN No.: 1477 MDL No.: MFCD00149875
Formel: N₃O₉Tm·xH₂O
Molecular Weight: 354.95 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149875
CAS-Nummer: 36548-87-5
EINECS: 239-069-9
UN: 1477
ADR: 5.1 III