Ethyl-4-chloracetoacetat ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Ethyl 4-chloro-3-oxobutanoate
A15641.0B
A15641.18
A15641.30
A15641.0BEA
297
EUR
A15641.0B
A15641.18
A15641.30
Ethyl-4-chloracetoacetat ≥97%
Ethyl-4-chloracetoacetat
Purity: 97%
CAS No.: 638-07-3
Molecular Formula: C6H9ClO3
Formula Weight: 164.59
Flash Point: 92°(198°F)
Refractive Index: 1.4520
Melting Point: -8°
Boiling Point: 220°
Beilstein Reference: 1761275
UN No.: 2922
MDL No.: MFCD00000939
Formel:
C₆H₉ClO₃ Molecular Weight: 164,59 g/mol Siedepunkt: 220 °C Schmelzpunkt: –8 °C Dichte: 1,212 Flammpunkt: 92 °C (198 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000939 CAS-Nummer: 638-07-3 EINECS: 211-317-0 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...