Ethyl-4-chloracetoacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl 4-chloro-3-oxobutanoate
M-Clarity™ Qualitätsstufe = MQ100
690295-1KG
690295-200KG
690295-25KG
SIAL690295-1KGEA
255
EUR
SIAL690295-1KG
SIAL690295-200KG
SIAL690295-25KG
Ethyl-4-chloracetoacetat, Sigma-Aldrich®
Ethyl-4-chloracetoacetat
Formel:
C₆H₉ClO₃ Molecular Weight: 164,59 g/mol |
MDL:
MFCD00000939 CAS-Nummer: 638-07-3 EINECS: 211-317-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...