Ethyl-4,4-difluoracetoacetat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-3-oxobutyrate
001566-1G
001566-5G
001566-25G
FLUO001566-1GEA
36.9
EUR
FLUO001566-1G
FLUO001566-5G
FLUO001566-25G
Ethyl-4,4-difluoracetoacetat
Ethyl-4,4-difluoracetoacetat
Formel:
C₆H₈F₂O₃ |
MDL:
MFCD00039277 CAS-Nummer: 352-24-9 EINECS: 206-519-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...