2,2-Difluoracetylchlorid

Lieferant: CHEMPUR

FL5184.5 FL5184.25
CHMPFL5184.5EA 0 EUR
CHMPFL5184.5 CHMPFL5184.25
2,2-Difluoracetylchlorid
2,2-Difluoracetylchlorid
Formel: C₂HClF₂O
CAS-Nummer: 381-72-6
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR