(2-Bromphenoxy)acetonitril

Lieferant: COMBI-BLOCKS

OT-1107-5G OT-1107-25G OT-1107-1G
COBBOT-1107-5GEA 204 EUR
COBBOT-1107-5G COBBOT-1107-25G COBBOT-1107-1G
(2-Bromphenoxy)acetonitril
(2-Bromphenoxy)acetonitril
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
MDL: MFCD00798567
CAS-Nummer: 90004-90-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR