(3-Bromphenoxy)acetonitril

Lieferant: COMBI-BLOCKS

HC-3218-5G HC-3218-1G
COBBHC-3218-5GEA 310 EUR
COBBHC-3218-5G COBBHC-3218-1G
(3-Bromphenoxy)acetonitril
(3-Bromphenoxy)acetonitril
Formel: C₈H₆BrNO
MDL: MFCD09906440
CAS-Nummer: 951918-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR