(4-Bromphenoxy)acetonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Bromophenoxy)acetonitrile

HC-3219-5G HC-3219-25G HC-3219-1G
COBBHC-3219-5GEA 335 EUR
COBBHC-3219-5G COBBHC-3219-25G COBBHC-3219-1G
(4-Bromphenoxy)acetonitril
(4-Bromphenoxy)acetonitril
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
MDL: MFCD00464958
CAS-Nummer: 39489-67-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR