(2,6-Dichlorphenoxy)acetonitril

Lieferant: Thermo Scientific

SPB02452.1 SPB02452.10 SPB02452.50
MAYBSPB02452.1EA 0 EUR
MAYBSPB02452.1 MAYBSPB02452.10 MAYBSPB02452.50
(2,6-Dichlorphenoxy)acetonitril
(2,6-Dichlorphenoxy)acetonitril
Formel: C₈H₅Cl₂NO
CAS-Nummer: 21244-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR