(2-Chlorphenoxy)acetonitril

Lieferant: Thermo Scientific

SPB01309.1 SPB01309.10 SPB01309.50
MAYBSPB01309.1EA 0 EUR
MAYBSPB01309.1 MAYBSPB01309.10 MAYBSPB01309.50
(2-Chlorphenoxy)acetonitril
(2-Chlorphenoxy)acetonitril
Formel: C₈H₆ClNO
Molecular Weight: 167,60 g/mol
MDL: MFCD00017331
CAS-Nummer: 43111-31-5
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR