(3-Chlorphenoxy)acetonitril

Lieferant: Thermo Scientific

SPB01310.1 SPB01310.10 SPB01310.50
MAYBSPB01310.1EA 0 EUR
MAYBSPB01310.1 MAYBSPB01310.10 MAYBSPB01310.50
(3-Chlorphenoxy)acetonitril
(3-Chlorphenoxy)acetonitril
Formel: C₈H₆ClNO
Molecular Weight: 167,60 g/mol
MDL: MFCD00017332
CAS-Nummer: 43111-32-6
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR