(4-Chlorphenoxy)acetonitril

Lieferant: Thermo Scientific

Synonyme: 2-(4-Chlorophenoxy)acetonitrile

RF03183.1 RF03183.10 RF03183.50
MAYBRF03183.1EA 0 EUR
MAYBRF03183.1 MAYBRF03183.10 MAYBRF03183.50
(4-Chlorphenoxy)acetonitril
(4-Chlorphenoxy)acetonitril
Formel: C₈H₆ClNO
Molecular Weight: 167,60 g/mol
MDL: MFCD00031535
CAS-Nummer: 3598-13-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR