1,1,1-Trifluor-2-butanon

Lieferant: CHEMPUR

FL1574.5 FL1574.25 001574-5G
CHMPFL1574.5EA 0 EUR
CHMPFL1574.5 CHMPFL1574.25 FLUO001574-5G
1,1,1-Trifluor-2-butanon
1,1,1-Trifluor-2-butanon
Formel: C₄H₅F₃O
Molecular Weight: 126,08 g/mol
MDL: MFCD00190641
CAS-Nummer: 381-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR