4,4,4-Trifluorbuttersäure
Lieferant: FLUOROCHEM
001553-1G
001553-5G
001553-25G
FLUO001553-1GEA
42.9
EUR
FLUO001553-1G
FLUO001553-5G
FLUO001553-25G
4,4,4-Trifluorbuttersäure
4,4,4-Trifluorbuttersäure
Formel:
C₄H₅F₃O₂ Molecular Weight: 142,08 g/mol |
MDL:
MFCD00077604 CAS-Nummer: 406-93-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...