4-Methyl-2-pentin ≥97%

Lieferant: Thermo Scientific
Gefahr

H53442.03 H53442.06 H53442.14
H53442.03EA 30.3 EUR
H53442.03 H53442.06 H53442.14
4-Methyl-2-pentin ≥97%
4-Methyl-2-pentyne

Purity: 97% CAS No.: 21020-27-9 Molecular Formula: C6H10 Formula Weight: 82.15 Flash Point: -12°(-10°F) Refractive Index: 1.407 Boiling Point: 71-73° UN No.: 3295 MDL No.: MFCD00041615

Formel: C₆H₁₀
Molecular Weight: 82,15 g/mol
Siedepunkt: 71…73 °C
Dichte: 0,711
Flammpunkt: –12 °C (10 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041615
CAS-Nummer: 21020-27-9
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR