Perfluor-1-butansulfonsäurefluorid ≥90%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride
B21322.06
B21322.14
B21322.22
B21322.06EA
28
EUR
B21322.06
B21322.14
B21322.22
Perfluor-1-butansulfonsäurefluorid ≥90%
Perfluor-1-butansulfonsäurefluorid
Purity: 90+%
CAS No.: 375-72-4
Molecular Formula: CF3(CF2)3SO2F
Formula Weight: 302.09
Flash Point: None
Refractive Index: 1.2810
Melting Point: -110°
Boiling Point: 65-66°
Beilstein Reference: 1813589
UN No.: 3265
MDL No.: MFCD00007422
Formel:
C₄F₁₀O₂S Molecular Weight: 302.09 g/mol Siedepunkt: 65…66 °C Schmelzpunkt: –110 °C Dichte: 1.750 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007422 CAS-Nummer: 375-72-4 EINECS: 206-792-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...